Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylparaben Sodium, NF, 94-102%, Spectrum™ Chemical
SDP

Catalog No. 18605924 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-924 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-605-924 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1457100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1457, 35285-69-9, C10H11NaO3

Spectrum™ Chemical Propylparaben Sodium, NF is used as an antimicrobial preservative. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 35285-69-9
Molecular Formula C10H11NaO3
Molecular Weight (g/mol) 202.19
InChI Key IXMINYBUNCWGER-UHFFFAOYSA-M
IUPAC Name sodium 4-(propoxycarbonyl)benzen-1-olate
SMILES [Na+].CCCOC(=O)C1=CC=C([O-])C=C1

Specifications

CAS Max % 1
pH 9.5 to 10.5
Packaging Poly Bottle
Quantity 100 g
Percent Purity 94 to 102%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Propylparaben Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.