accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CMgCl

CAS RN 35293-35-7

IUPAC Name: chloro(2-methyl-2-phenylpropyl)magnesium
Synonyms: chloro(2-methyl-2-phenylpropyl)magnesium
Molecular Weight (g/mol): 192.97
Molecular Formula: C10H13ClMg
InChi Key: BXKYFONHSSFLKR-UHFFFAOYSA-M
SMILES: CC(C)(C[Mg]Cl)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)