Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich P-Tolyltrichlorosilane

Catalog No. 41935425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-935-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 41-935-425G Supplier Sigma Aldrich Supplier No. 41935425G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Self-assembled monolayers (SAMs) of p-tolyltrichlorosilane (TTCS) were formed on flexible polymer substrates. p-Tolyltrichlorosilane copolymerized with methylphenyldichlorosilane was grafted on silicon to introduce a structural defect.

Specifications

CAS 35293-35-7
Density 0.766 g/mL at 25°C
Boiling Point 34.6°C
Molecular Formula C10H13ClMg
Linear Formula C6H5C(CH3)2CH2MgCl
Refractive Index n20/D 1.5240
MDL Number MFCD00075586
Quantity 25 g
Molecular Weight (g/mol) 192.97 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.