accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 353-42-4

IUPAC Name: methoxymethane; trifluoroborane
Synonyms: boron trifluoride; methyl ether
Molecular Weight (g/mol): 113.87
Molecular Formula: C2H6BF3O
InChi Key: KTPWETRNSUKEME-UHFFFAOYSA-N
SMILES: COC.FB(F)F
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)