accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 353-43-5

IUPAC Name: (methylsulfanyl)methane; trifluoroborane
Synonyms: boron trifluoride; dimethyl sulfide
Molecular Weight (g/mol): 129.94
Molecular Formula: C2H6BF3S
InChi Key: BRWZPVRDOUWXKE-UHFFFAOYSA-N
SMILES: CSC.FB(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results