Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trihexylsilane

Catalog No. 27614625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-614-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 27-614-625G Supplier Sigma Aldrich Supplier No. 27614625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for: Enantioselective Si-H insertionRegio- and stereoselective oxidation into silanolsAsymmetric carbenoid insertion of diazoacetates into the silicon-hydrogen bondHydrosilylation of carbon dioxideCatalyst for hydrodefluorination leading to the synthesis of hydrocarbons

Specifications

CAS 353-43-5
Density 1.235 g/mL at 25°C
Molecular Formula C2H6BF3S
Refractive Index n20/D 1.448
Linear Formula (CH3)2S -+ BF3
MDL Number MFCD00013193
Quantity 25 g
Synonym NSC 139856
Molecular Weight (g/mol) 284.6 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.