accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3531-19-9

IUPAC Name: 2-chloro-4,6-dinitroaniline
Synonyms: 2-chloro-4,6-dinitroaniline
Molecular Weight (g/mol): 217.57
Molecular Formula: C6H4ClN3O4
InChi Key: LHRIICYSGQGXSX-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)