Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-2,4-dinitroaniline 98.0+%, TCI America™
SDP

Catalog No. C065725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C065725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C065725G Supplier TCI America Supplier No. C065725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3531-19-9
Molecular Formula C6H4ClN3O4
Molecular Weight (g/mol) 217.57
MDL Number MFCD00007147
InChI Key LHRIICYSGQGXSX-UHFFFAOYSA-N
Synonym 6-chloro-2,4-dinitroaniline, 2-chloro-4,6-dinitrophenyl amine, benzenamine, 2-chloro-4,6-dinitro, ccris 3112, aniline, 6-chloro-2,4-dinitro, 6-chloro-2,4-dinitrophenylamine, acmc-209ig0, 2,4-dinitro-6-chloroaniline, 3-12-00-01718 beilstein handbook reference, aronis23887
PubChem CID 19052
IUPAC Name 2-chloro-4,6-dinitroaniline
SMILES NC1=C(Cl)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 158°C
Quantity 25 g
UN Number 2811
Formula Weight 217.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-2,4-dinitroaniline
EINECSNumber (3)-0422
RTECSNumber BX1226000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.