accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 3531-24-6

IUPAC Name: 3,3-diphenylprop-2-enenitrile
Synonyms: 3,3-diphenylprop-2-enenitrile
Molecular Weight (g/mol): 205.26
Molecular Formula: C15H11N
InChi Key: RDGWQFSLTSPRBG-UHFFFAOYSA-N
SMILES: N#CC=C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results