Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-(Diphenyl)acrylonitrile 98.0+%, TCI America™
SDP

Catalog No. D4873200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4873200MG 200 mg
D48731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4873200MG Supplier TCI America Supplier No. D4873200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3531-24-6
Molecular Formula C15H11N
Molecular Weight (g/mol) 205.26
MDL Number MFCD00045630
InChI Key RDGWQFSLTSPRBG-UHFFFAOYSA-N
PubChem CID 274352
IUPAC Name 3,3-diphenylprop-2-enenitrile
SMILES N#CC=C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 45°C
Color Yellow
Boiling Point 131°C
Quantity 200 mg
Formula Weight 205.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3-(Diphenyl)acrylonitrile
EINECSNumber (4)-0918
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.