accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35335-16-1

IUPAC Name: 1,4-dibromo-2,5-bis(bromomethyl)benzene
Synonyms: 1,4-dibromo-2,5-bis(bromomethyl)benzene
Molecular Weight (g/mol): 421.75
Molecular Formula: C8H6Br4
InChi Key: UOCMTKLYRROETA-UHFFFAOYSA-N
SMILES: BrCC1=CC(Br)=C(CBr)C=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results