Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dibromo-2,5-bis(bromomethyl)benzene 97.0+%, TCI America™
SDP

Catalog No. D39615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39615G 5 g
D39611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39615G Supplier TCI America Supplier No. D39615G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35335-16-1
Molecular Formula C8H6Br4
Molecular Weight (g/mol) 421.75
MDL Number MFCD03411745
InChI Key UOCMTKLYRROETA-UHFFFAOYSA-N
Synonym alpha,alpha,2,5-Tetrabromo-p-xylene
PubChem CID 640222
IUPAC Name 1,4-dibromo-2,5-bis(bromomethyl)benzene
SMILES BrCC1=CC(Br)=C(CBr)C=C1Br

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 421.75
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromo-2,5-bis(bromomethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.