accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3535-84-0

IUPAC Name: sulfanylformonitrile thallium
Synonyms: thallium thiocyanate
Molecular Weight (g/mol): 263.47
Molecular Formula: CHNSTl
InChi Key: BEORFWVICVXJAG-UHFFFAOYSA-N
SMILES: [Tl].SC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results