Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium(I) thiocyanate, 98%, Thermo Scientific™

Catalog No. AAA1697214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1697214 25 g
AAA1697206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1697214 Supplier Thermo Scientific Chemicals Supplier No. A1697214

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3535-84-0
Molecular Formula CNSTl
Molecular Weight (g/mol) 262.458
MDL Number MFCD00049608
InChI Key JSSGUHHJIJMEQF-UHFFFAOYSA-M
Synonym thallium thiocyanate, thallium 1+ thiocyanate, thiocyanate ion thallane, thiocyanate ion thallium hydride, thiocyanic acid, thallium 1+ salt
PubChem CID 3082002
IUPAC Name thallium(1+);thiocyanate
SMILES C(#N)[S-].[Tl+]

Specifications

Quantity 25 g
UN Number UN1707
Formula Weight 262.46
Percent Purity 98%
Chemical Name or Material Thallium(I) thiocyanate
Hazard Category Danger
Hazard Statement GHS H Statement
H300-H330-H373-H312
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.