accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaCH3NCH3CNCH3H3COHSOOOSOOOCH3NCH3CNCH3H3COHSOOOSOOO

CAS RN 3536-49-0

IUPAC Name: calcium bis(bis[4-(diethylamino)phenyl](5-hydroxy-2,4-disulfonatophenyl)methylium)
Synonyms: patent blue V
Molecular Weight (g/mol): 1159.42
Molecular Formula: C54H62CaN4O14S4
InChi Key: RTMBGDBBDQKNNZ-UHFFFAOYSA-L
SMILES: [Ca++].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC(O)=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O.CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC(O)=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results