Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylene Blue V, MP Biomedicals

Catalog No. ICN219994 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219994 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219994 Supplier MP Biomedicals, Inc Supplier No. 0521999410

Chemical Identifiers

CAS 3536-49-0
Molecular Formula C54H62CaN4O14S4
Molecular Weight (g/mol) 1159.422
InChI Key RTMBGDBBDQKNNZ-UHFFFAOYSA-L
PubChem CID 77073
IUPAC Name calcium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonate
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC(=C(C=C3S(=O)(=O)[O-])S(=O)(=O)[O-])O.CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC(=C(C=C3S(=O)(=O)[O-])S(=O)(=O)[O-])O.[Ca+2]

Specifications

Quantity 10 g
Chemical Name or Material Xylene Blue V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.