accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFOHNOHNCl

CAS RN 35367-38-5

IUPAC Name: 1-(4-chlorophenyl)-3-(2,6-difluorobenzoyl)urea
Synonyms: diflubenzuron difluron
Molecular Weight (g/mol): 310.68
Molecular Formula: C14H9ClF2N2O2
InChi Key: QQQYTWIFVNKMRW-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1C(=O)NC(=O)NC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results