accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH3C(S)(S)(R)OH(S)(S)HOH3C(S)(S)(R)OH(S)(S)HO

CAS RN 35380-71-3

IUPAC Name: bis((1S,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol) hydrate
Synonyms: bis(estradiol) hydrate
Molecular Weight (g/mol): 562.79
Molecular Formula: C36H50O5
InChi Key: ZVVGLAMWAQMPDR-WVEWYJOQSA-N
SMILES: O.C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O.C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O
Grade 
  • (11)
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (8)
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (11)
  • (8)
  • (3)
  • (3)
  • (11)
  • (8)
  • (3)
  • (8)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)