Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estradiol, Hemihydrate, Soy Base, Micronized, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18602451 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-451 500 g Amber Glass Bottle
18-602-449 25 g Amber Glass Bottle
18-602-450 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-451 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E2918500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

E2918, 35380-71-3, C18H24O2·1/2H2O

Spectrum™ Chemical Estradiol, Hemihydrate, Soy Base, Micronized, USP

Chemical Identifiers

CAS 35380-71-3
Molecular Formula C36H50O5
Molecular Weight (g/mol) 562.79
InChI Key ZVVGLAMWAQMPDR-WVEWYJOQSA-N
IUPAC Name bis((1S,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol) hydrate
SMILES O.C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O.C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O

Specifications

CAS Max % 100%
Melting Range 173°-179°C
Packaging Amber Glass Bottle
Quantity 500 g
Specific Rotation +76° to +83°
Percent Purity 97 to 103%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Estradiol
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.