CAS RN 354-51-8
CAS RN 354-51-8
IUPAC Name:
1,2-dibromo-1-chloro-1,2,2-trifluoroethane
Synonyms:
1,2-dibromo-1-chloro-1,2,2-trifluoroethane
Molecular Weight (g/mol):
276.28
Molecular Formula:
C2Br2ClF3
InChi Key:
OVZATIUQXBLIQT-UHFFFAOYNA-N
SMILES:
FC(F)(Br)C(F)(Cl)Br
1
–
1
of
1
results
1
1,2-Dibromo-1-chloro-1,2,2-trifluoroethane, TCI America™
CAS: 354-51-8 Molecular Formula: C2Br2ClF3 Molecular Weight (g/mol): 276.275 MDL Number: MFCD00039316 InChI Key: OVZATIUQXBLIQT-UHFFFAOYSA-N Synonym: Flon-113B-2 PubChem CID: 9634 IUPAC Name: 1,2-dibromo-1-chloro-1,2,2-trifluoroethane SMILES: C(C(F)(Cl)Br)(F)(F)Br