Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromo-1-chloro-1,2,2-trifluoroethane, TCI America™
SDP

Catalog No. D222025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D222025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D222025G Supplier TCI America Supplier No. D222025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 354-51-8
Molecular Formula C2Br2ClF3
Molecular Weight (g/mol) 276.275
MDL Number MFCD00039316
InChI Key OVZATIUQXBLIQT-UHFFFAOYSA-N
Synonym Flon-113B-2
PubChem CID 9634
IUPAC Name 1,2-dibromo-1-chloro-1,2,2-trifluoroethane
SMILES C(C(F)(Cl)Br)(F)(F)Br

Specifications

Color Colorless
Boiling Point 93°C
Quantity 25 g
Formula Weight 276.28
Physical Form Liquid
Chemical Name or Material 1,2-Dibromo-1-chloro-1,2,2-trifluoroethane
RTECSNumber KH9300000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.