accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNOOHSOOHOSOOHOAsOOHHONaNa(E/Z)

CAS RN 3547-38-4

IUPAC Name: 3-[2-(2-arsonophenyl)hydrazin-1-ylidene]-5-hydroxy-4-oxo-3,4-dihydronaphthalene-2,7-disulfonic acid disodium
Synonyms: 3-[2-(2-arsonophenyl)hydrazin-1-ylidene]-5-hydroxy-4-oxonaphthalene-2,7-disulfonic acid disodium
Molecular Weight (g/mol): 594.3
Molecular Formula: C16H13AsN2Na2O11S2
InChi Key: HLLSKRQLMZTAFK-UHFFFAOYSA-N
SMILES: [Na].[Na].OC1=C2C(=O)C(=NNC3=CC=CC=C3[As](O)(O)=O)C(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results