Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenazo I Hydrate 90.0+%, TCI America™
SDP

Catalog No. D05781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D05781G Supplier TCI America Supplier No. D05781G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3547-38-4
Molecular Formula C16H13AsN2Na2O11S2
Molecular Weight (g/mol) 594.304
MDL Number MFCD00066725
InChI Key HLLSKRQLMZTAFK-AXHWYJOWSA-N
Synonym 2-Arsonophenylazochromotropic Acid Sodium Salt, 2-(1,8-Dihydroxy-3,6-disulfo-2-naphthylazo)benzenearsonic Acid Sodium Salt, Neothorin
PubChem CID 131878335
IUPAC Name (3E)-3-[(2-arsonophenyl)hydrazinylidene]-5-hydroxy-4-oxonaphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O.[Na].[Na]

Specifications

Quantity 1 g
UN Number 3465
Formula Weight 614.27
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Arsenazo I Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.