accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 355-43-1

IUPAC Name: 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
Synonyms: hexane, tridecafluoro-6-iodo-
Molecular Weight (g/mol): 445.95
Molecular Formula: C6F13I
InChi Key: BULLJMKUVKYZDJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Boiling Point 
  • (2)
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)