Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoro-1-iodohexane, 98+%, stab. with copper

Catalog No. AAB2095830
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2095830 250 g
AAB2095809 10 g
AAB2095818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2095830 Supplier Thermo Scientific Chemicals Supplier No. B2095830
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 355-43-1 | C6F13I | 445.95 g/mol

It is used as an initiator in the presence of fac-[Ir(ppy) 3 for the photoredox catalyst activated CRP of methacrylates in visible light-induced controlled radical polymerization of methacrylates with perfluoroalkyl iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an initiator in the presence of fac-[Ir(ppy) 3 for the photoredox catalyst activated CRP of methacrylates in visible light-induced controlled radical polymerization of methacrylates with perfluoroalkyl iodide.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-43-1
Molecular Formula C6F13I
Molecular Weight (g/mol) 445.95
MDL Number MFCD00001063
InChI Key BULLJMKUVKYZDJ-UHFFFAOYSA-N
Synonym perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo
PubChem CID 67733
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
SMILES C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Melting Point -45°C
Density 2.063
Boiling Point 117°C
Refractive Index 1.329
Quantity 250 g
Beilstein 1802118
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 445.95
Percent Purity ≥98%
Chemical Name or Material Perfluoro-1-iodohexane, stabilized with copper
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-586-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.