accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONOCH3N

CAS RN 3551-55-1

IUPAC Name: 2,4-dimethoxypyrimidine
Synonyms: 2,4-dimethoxypyrimidine
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: KEVRHVMWBKFGLO-UHFFFAOYSA-N
SMILES: COC1=CC=NC(OC)=N1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)