accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 355386-94-6

IUPAC Name: (quinolin-5-yl)boronic acid
Synonyms: quinolin-5-ylboronic acid
Molecular Weight (g/mol): 172.98
Molecular Formula: C9H8BNO2
InChi Key: NWIJBOCPTGHGIK-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=NC2=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results