Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinoline-5-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. Q0087200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0087200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q0087200MG Supplier TCI America Supplier No. Q0087200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 355386-94-6
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.978
MDL Number MFCD03095058
InChI Key NWIJBOCPTGHGIK-UHFFFAOYSA-N
Synonym quinoline-5-boronic acid, 5-quinolineboronic acid, quinolin-5-yl-5-boronic acid, 5-quinolinylboronic acid, boronic acid, 5-quinolinyl, quinolin-5-yl boronic acid, 5-quinolineboronicacid, 5-boronoquinoline, pubchem1879, 5-quinolylboronic acid
PubChem CID 5153389
IUPAC Name quinolin-5-ylboronic acid
SMILES B(C1=C2C=CC=NC2=CC=C1)(O)O

Specifications

Quantity 200 mg
Formula Weight 172.98
Physical Form Crystalline Powder
Chemical Name or Material Quinoline-5-boronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.