accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3556-83-0

IUPAC Name: methyl 3-methoxy-4-methylbenzoate
Synonyms: methyl 3-methoxy-4-methylbenzoate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: LLEXCSBUSVRBCA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C)C(OC)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)