accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35598-05-1

IUPAC Name: methyl 4-methoxy-2-methylbenzoate
Synonyms: methyl 4-methoxy-2-methylbenzoate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: OGYAVWKYDVBIMW-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C=C(OC)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results