accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOCH3FFFF

CAS RN 356-36-5

IUPAC Name: 1,4-dimethyl 2,2,3,3-tetrafluorobutanedioate
Synonyms: 1,4-dimethyl 2,2,3,3-tetrafluorobutanedioate
Molecular Weight (g/mol): 218.1
Molecular Formula: C6H6F4O4
InChi Key: RMXAYQBUNVSEPG-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results