Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Tetrafluorosuccinate 95.0+%, TCI America™
SDP

Catalog No. D35885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35885G 5 g
D35881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35885G Supplier TCI America Supplier No. D35885G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 356-36-5
Molecular Formula C6H6F4O4
Molecular Weight (g/mol) 218.104
MDL Number MFCD00043947
InChI Key RMXAYQBUNVSEPG-UHFFFAOYSA-N
Synonym Dimethyl Perfluorosuccinate, Perfluorosuccinic Acid Dimethyl Ester, Tetrafluorosuccinic Acid Dimethyl Ester, Dimethyl Tetrafluorobutanedioate, Tetrafluorobutanedioic Acid Dimethyl Ester
PubChem CID 2737124
IUPAC Name dimethyl 2,2,3,3-tetrafluorobutanedioate
SMILES COC(=O)C(C(C(=O)OC)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 174°C
Quantity 5 g
Formula Weight 218.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Tetrafluorosuccinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.