accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFOOH2C

CAS RN 356-86-5

IUPAC Name: 2,2,3,3,3-pentafluoropropyl prop-2-enoate
Synonyms: 2,2,3,3,3-pentafluoropropyl prop-2-enoate
Molecular Weight (g/mol): 204.1
Molecular Formula: C6H5F5O2
InChi Key: JDVGNKIUXZQTFD-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)COC(=O)C=C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)