Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,3-Pentafluoropropyl Acrylate (stabilized with TBC) 98.0+%, TCI America™
SDP

Catalog No. P23525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23525G Supplier TCI America Supplier No. P23525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 356-86-5
Molecular Formula C6H5F5O2
Molecular Weight (g/mol) 204.10
MDL Number MFCD00039257
InChI Key JDVGNKIUXZQTFD-UHFFFAOYSA-N
Synonym 2,2,3,3,3-pentafluoropropyl acrylate, 1h,1h-pentafluoropropyl acrylate, 2-propenoic acid, 2,2,3,3,3-pentafluoropropyl ester, acmc-20aokp, acrylic acid=2,2,3,3,3-pentafluoropropyl ester, acrylic acid 2,2,3,3,3-pentafluoropropyl ester, 2-propenoic acid,2,2,3,3,3-pentafluoropropyl ester, 2,2,3,3,3-pentafluoropropyl acrylate stabilized with tbc, 2,2,3,3,3-pentafluoropropyl acrylate, contains 100 ppm 4-tert-butylcatechol as inhibitor
PubChem CID 67744
IUPAC Name 2,2,3,3,3-pentafluoropropyl prop-2-enoate
SMILES FC(F)(F)C(F)(F)COC(=O)C=C

Specifications

Color Colorless
Boiling Point 50°C
UN Number 3272
Quantity 5 g
Formula Weight 204.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,3-Pentafluoropropyl Acrylate (stabilized with TBC)
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.