accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNH2NHO(E)

CAS RN 35613-84-4

IUPAC Name: (E)-N'-hydroxy-2-methylpropanimidamide
Synonyms: (E)-N'-hydroxy-2-methylpropanimidamide
Molecular Weight (g/mol): 102.14
Molecular Formula: C4H10N2O
InChi Key: JHRDEHLFNLLCQS-UHFFFAOYSA-N
SMILES: CC(C)C(\N)=N/O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results