Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyramidoxime, 96%

Catalog No. AAH5089203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5089203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5089203 Supplier Thermo Scientific Chemicals Supplier No. H5089203

missing translation for 'validMainframeMsg'

CAS: 35613-84-4 | C4H10N2O | 102.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35613-84-4
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
MDL Number MFCD03426251,MFCD08705910,MFCD00464225
InChI Key JHRDEHLFNLLCQS-UHFFFAOYSA-N
Synonym z-n'-hydroxyisobutyrimidamide, 1z-n'-hydroxy-2-methylpropanimidamide, n-hydroxyisobutyrimidamide, n-hydroxy-2-methylpropanimidamide, z-n'-hydroxy-2-methylpropimidamide, n-hydroxyisobutyramidine, z-n'-hydroxy-2-methylpropanimidamide, isobutyramidoxime 250mg, n/'-hydroxy-2-methylpropanimidamide, propanimidamide, n-hydroxy-2-methyl
PubChem CID 9582835
IUPAC Name N'-hydroxy-2-methylpropanimidamide
SMILES CC(C)C(\N)=N/O

Specifications

Melting Point 57°C to 65°C
Quantity 1 g
Formula Weight 102.14
Percent Purity 96%
Chemical Name or Material Isobutyramidoxime
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.