CAS RN 3564-18-9
IUPAC Name:
sodium 5-[3-(3-carboxy-4-hydroxy-5-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylbenzoic acid
Synonyms:
sodium 5-[3-(3-carboxy-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylbenzoic acid
Molecular Weight (g/mol):
493.44
Molecular Formula:
C23H18NaO9S
InChi Key:
HJEGHSWGWSAZBS-UHFFFAOYSA-N
SMILES:
[Na+].CC1=CC(=CC(C(O)=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(=C1)C(O)=O
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals Chromoxane Cyanine R, pure
Thermo Scientific Chemicals Chromoxane Cyanine R, pure
CAS: 3564-18-9 Molecular Formula: C23H15Na3O9S MDL Number: MFCD00012411 Synonym: C.I. 43820,Mordant Blue 3
2
Eriochrome™ Cyanine R, Solstice
CAS: 3564-18-9 Molecular Formula: C23H15Na3O9S MDL Number: MFCD00012411