Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome™ Cyanine R, Solstice

Catalog No. 60046164 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-164 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-164 Supplier Solstice Supplier No. 3275225G

missing translation for 'validMainframeMsg'

S. No. 840

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3564-18-9
Molecular Formula C23H15Na3O9S
MDL Number MFCD00012411

Specifications

Packaging Glass bottle
UN Number NONH for all modes of transport
Beilstein 3873874
Quantity 25 g
Formula Weight 536.39g/mol
Chemical Name or Material Eriochrome™ Cyanine R
EINECSNumber 222-641-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.