accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35657-36-4

IUPAC Name: amino 4-nitrobenzoate
Synonyms: amino 4-nitrobenzoate
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H6N2O4
InChi Key: VTKRSTQMGNRMHL-UHFFFAOYSA-N
SMILES: NOC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)