Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(4-Nitrobenzoyl)hydroxylamine 97.0+%, TCI America™
SDP

Catalog No. N10135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10135G 5 g
N10131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10135G Supplier TCI America Supplier No. N10135G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35657-36-4
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.14
MDL Number MFCD11976095
InChI Key VTKRSTQMGNRMHL-UHFFFAOYSA-N
PubChem CID 426419
IUPAC Name amino 4-nitrobenzoate
SMILES NOC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 5 g
Formula Weight 182.14
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material O-(4-Nitrobenzoyl)hydroxylamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.