accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35661-51-9

IUPAC Name: [(9H-fluoren-9-yl)methoxy]carbohydrazide
Synonyms: 9H-fluoren-9-ylmethoxycarbohydrazide
Molecular Weight (g/mol): 254.29
Molecular Formula: C15H14N2O2
InChi Key: YGCGPEUVGHDMLO-UHFFFAOYSA-N
SMILES: NNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results