Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenylmethyl Carbazate 98.0+%, TCI America™
SDP

Catalog No. F08725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08725G 5 g
F087225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08725G Supplier TCI America Supplier No. F08725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35661-51-9
Molecular Formula C15H14N2O2
Molecular Weight (g/mol) 254.29
MDL Number MFCD00239419
InChI Key YGCGPEUVGHDMLO-UHFFFAOYSA-N
Synonym Carbazic Acid 9-Fluorenylmethyl Ester, Fmoc-hydrazine, [(9H-Fluoren-9-ylmethoxy)carbonyl]hydrazine
PubChem CID 161883
IUPAC Name [(9H-fluoren-9-yl)methoxy]carbohydrazide
SMILES NNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 171°C
Color White
Quantity 5 g
Formula Weight 254.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenylmethyl Carbazate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.