accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3569-18-4

IUPAC Name: (3R)-3-(4-carboxylatophenyl)-1,1,3-trimethyl-2,3-dihydro-1H-indene-5-carboxylate
Synonyms: (3R)-3-(4-carboxylatophenyl)-1,1,3-trimethyl-2H-indene-5-carboxylate
Molecular Weight (g/mol): 322.36
Molecular Formula: C20H18O4
InChi Key: CDZMWAHBQLPCHD-HXUWFJFHSA-L
SMILES: CC1(C)C[C@@](C)(C2=CC(=CC=C12)C([O-])=O)C1=CC=C(C=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results