Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Carboxyphenyl)-2,3-dihydro-1,1,3-trimethyl-1H-indene-5-carboxylic Acid, Thermo Scientific™

Catalog No. AC404131000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC404131000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC404131000 Supplier Thermo Scientific Chemicals Supplier No. 404131000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3569-18-4
Molecular Formula C20H20O4
Molecular Weight (g/mol) 324.376
InChI Key CDZMWAHBQLPCHD-UHFFFAOYSA-N
Synonym 1,1,3-trimethyl-3-phenylindan-4',5-dicarboxylic acid, 3-4-carboxyphenyl-1,1,3-trimethylindane-5-carboxylic acid, 3-4-carboxyphenyl-1,1,3-trimethyl-2h-indene-5-carboxylic acid, 1h-indene-5-carboxylic acid, 3-4-carboxyphenyl-2,3-dihydro-1,1,3-trimethyl, 1h-indene-5-carboxylicacid, 3-4-carboxyphenyl-2,3-dihydro-1,1,3-trimethyl, 3-4-carboxyphenyl-2,3-dihydro-1,1,3-trimethyl-1h-indene-5-carboxylic acid, 1,1,3-trimethyl 3-4-carboxyphenyl indan-5-carboxylic acid, 3-phenyl-1,1,3-trimethylindan-4',5-dicarboxylic acid, 3-4-carboxyphenyl-1,1,3-trimethyl-2,3-dihydro-1h-indene-5-carboxylic acid
PubChem CID 107125
IUPAC Name 3-(4-carboxyphenyl)-1,1,3-trimethyl-2H-indene-5-carboxylic acid
SMILES CC1(CC(C2=C1C=CC(=C2)C(=O)O)(C)C3=CC=C(C=C3)C(=O)O)C

Specifications

Quantity 100 g
Formula Weight 324.38
Chemical Name or Material 3-(4-Carboxyphenyl)-2,3-dihydro-1,1,3-trimethyl-1H-indene-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.