accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 357271-55-7

OOOOHOONNH(S)CH3CH3H3C

CAS RN 357271-55-7

IUPAC Name: (3S)-4-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: (3S)-4-[1-(tert-butoxycarbonyl)indol-3-yl]-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol): 540.62
Molecular Formula: C32H32N2O6
InChi Key: HJKUZJNYQMKUEN-NRFANRHFSA-N
SMILES: CC(C)(C)OC(=O)N1C=C(C[C@@H](CC(O)=O)NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C2=CC=CC=C12
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)