accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3574-96-7

ClHNN

CAS RN 3574-96-7

IUPAC Name: 2-(2-chlorophenyl)-1H-1,3-benzodiazole
Synonyms: 2-(2-chlorophenyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 228.68
Molecular Formula: C13H9ClN2
InChi Key: OILIYWFQRJOPAI-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results