Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-chlorophenyl)benzimidazole, 97%, Thermo Scientific™

Catalog No. AC439040250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439040250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC439040250 Supplier Thermo Scientific Chemicals Supplier No. 439040250

Chemical Identifiers

CAS 3574-96-7
Molecular Formula C13H9ClN2
Molecular Weight (g/mol) 228.68
InChI Key OILIYWFQRJOPAI-UHFFFAOYSA-N
Synonym chlorfenazole, 2-2-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-benzimidazole, chlorfenazole iso, 1h-benzimidazole, 2-2-chlorophenyl, 2-2-chlorophenyl-1h-benzoimidazole, 2-2-chlorophenyl-1h-benzo d imidazole, unii-rt668w2svp, 2-o-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-1,3-benzodiazole
PubChem CID 77123
IUPAC Name 2-(2-chlorophenyl)-1H-benzimidazole
SMILES C1=CC=C(C(=C1)C2=NC3=CC=CC=C3N2)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 223.2°C to 234.7°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Formula Weight 228.68
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-chlorophenyl)benzimidazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.