accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35778-58-6

IUPAC Name: (5Z)-5-{[4-(diethylamino)phenyl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms: (5Z)-5-{[4-(diethylamino)phenyl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 292.42
Molecular Formula: C14H16N2OS2
InChi Key: CWQLQYNQWCTDQF-XFXZXTDPSA-N
SMILES: CCN(CC)C1=CC=C(\C=C2/SC(=S)NC2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results