Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-[4-(Diethylamino)benzylidene]rhodanine 98.0+%, TCI America™
SDP

Catalog No. D04645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04645G 5 g
D04641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D04645G Supplier TCI America Supplier No. D04645G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35778-58-6
Molecular Formula C14H16N2OS2
Molecular Weight (g/mol) 292.42
MDL Number MFCD00193758,MFCD00193758
InChI Key CWQLQYNQWCTDQF-XFXZXTDPSA-N
PubChem CID 3034543
IUPAC Name (5Z)-5-{[4-(diethylamino)phenyl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CCN(CC)C1=CC=C(\C=C2/SC(=S)NC2=O)C=C1

Specifications

Color Purple-Red
Quantity 5 g
Synonym p-diethylaminobenzylidene rhodanine, 5-4-diethylamino benzylidene rhodanine, 4-thiazolidinone, 5-4-diethylamino phenyl methylene-2-thioxo, acmc-209ij5, 5-4-diethylamino phenyl methylidene-2-sulfanylidene-1,3-thiazolidin-4-one, 4-thiazolidinone,5-4-diethylamino phenyl methylene-2-thioxo
Formula Weight 292.42
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-[4-(Diethylamino)benzylidene]rhodanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.