accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONHOHO(E)

CAS RN 35842-45-6

IUPAC Name: 2-[(2E)-2-methylbut-2-enamido]acetic acid
Synonyms: [(2E)-2-methylbut-2-enamido]acetic acid
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: WRUSVQOKJIDBLP-HWKANZROSA-N
SMILES: C\C=C(/C)C(=O)NCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results